Scoop, 1 ml | Avantor
Scoop, 1 ml
Scoop, 1 mL
Catalog # 470153-822
Supplier:   Bel-Art
Fournisseur #:  F36721-0100_SE
Scoop, 1 ml
Catalog # 470153-822
Supplier:  Bel-Art
Scoop, 1 ml
Scoop, 1 mL
Price
$526.65
Log in pour la disponibilité
Pack of 200
Product Details & Documents
Small sampling scoop made from white high-impact polystyrene plastic.
Ideal for retrieving small, specific volume samples of powders or similar materials.
Spécifications
  • Désignation:
    Scoop, 1 mL