Mini Scoop, 0.25 ml | Avantor
Mini Scoop, 0.25 ml
Scoop, .25 g
Catalog # 470153-826
Supplier:   Bel-Art
Fournisseur #:  F36721-0025_SE
Mini Scoop, 0.25 ml
Catalog # 470153-826
Supplier:  Bel-Art
Mini Scoop, 0.25 ml
Scoop, .25 g
Price
$3.20
Log in pour la disponibilité
Each Pack of 25
Product Details & Documents
MIni sampling scoop made from white high-impact polystyrene plastic.
iIeal for retrieving small, specific volume samples of powders or similar materials.
Spécifications
  • Désignation:
    Scoop, .25 g