Scissors
20.3 cm (8")
Catalog # 470101-894
Supplier: EATON OFFICE SUPPLY
Fournisseur #: ACM10572
Jump to:
Product Details & Documents
Stainless Steel
- Rounded safety tip blades
- Comfort-contoured polypropylene handles
Caution: Contains functional sharp edges or points.
Spécifications
- Longueur:20.3 cm (8")