ContactUsLinkComponent
Scientific Method Problem Solving Kit
Catalog # 470002-462
Supplier:  Lab-Aids
undefined
Scientific Method Problem Solving Kit
Catalog # 470002-462
Supplier:  Lab-Aids
Supplier Number:  100-A
Specifications
  • Description:
    Scientific Method Problem-Solving Kit
  • No. of Lab Groups:
    15
  • Cat. no.:
    470002-462
  • Supplier No.:
    100-A
Specifications
About this item
Easy-To-Understand, Practical Application Of Solving A Problem
  • Materials For Up To 30 Students
  • Form And Test A Hypothesis Using A Chemical Reaction
  • Teachers Guide Included
Using observation and analysis of color changes in solutions with bromothymol blue indicator, students will employ the scientific method of defining a problem. They will collect data, formulate a hypothesis, and test the hypothesis, using both experimental and control data to conclude that a color change is usually evidence of a chemical reaction.