Ward's® pH Buffer Kit | Avantor
Ward's® pH Buffer Kit
Ward's Chemistry pH Buffer Kit, Kit, 4/7/10
Catalog # 470233-572
Supplier:   Ward's Science
Fournisseur #:  BB0608-KIT
Ward's® pH Buffer Kit
Catalog # 470233-572
Supplier:  Ward's Science
CAS Number:  
Ward's® pH Buffer Kit
Ward's® pH Buffer Kit
Ward's Chemistry pH Buffer Kit, Kit, 4/7/10
CAS Number:  
Price
$104.20
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Includes 500 ml each of buffer solution pH 10, pH 4, and pH 7. Also includes 100 ml of electrode storage solution, KCl.
  • Aligned with GLOBE protocols
Shelf life: If stored in unopened bottles, then it is one year from receipt. Once opened it should be used within six months of first use.
Spécifications
Spécifications
  • Désignation:
    Ward's Chemistry pH Buffer Kit
  • Taille:
    Kit
  • pH:
    4/7/10