Sand Dollar
Catalog # 470000-906
Supplier: Avantor
Specifications
- Description:Sand Dollar, Preserved, Jar of 10
- Cat. no.:470000-906
- Supplier No.:WARD470000-906
Specifications
About this item
Formalin Preserved
Echinarachnius. It displays the flattened radial symmetry characteristic of this order. Size: 2– 3" dia.