SAFE-T Measures® Retractable
Retractable tape measure, Safe-T, 3 m (10 ft.)
Catalog # 470046-224
Supplier: American Scientific
Fournisseur #: 3968-00
Jump to:
Product Details & Documents
Locks and holds into position and automatically retracts at the touch of a button.
Spécifications
- Désignation:Retractable tape measure, Safe-T
- Longueur:3 m (10 ft.)