Saccharomyces, Budding Cells Slide
Saccharomyces, Budding Cells, Whole mount, Harris Hematoxylin and Orange-G
Catalog # 470176-888
Supplier: Avantor
Jump to:
Product Details & Documents
Fungal Processes
- Identifying features clearly distinguishable
- Rigorous quality control standards
Budding yeast cells stained for nuclei.
Spécifications
- Désignation:Saccharomyces, Budding Cells
- Stain:Harris Hematoxylin and Orange-G
- Orientation:Whole mount