Assorted Rubber Stoppers
 :   Plasticoid
Assorted Rubber Stoppers
Assorted Rubber Stoppers
Assorted Rubber Stoppers
Solid, 1-hole and 2-hole stoppers in assorted sizes.
  • Superior grade of rubber
  • Soft, resilent, and durable
  • Low sulfur content
  • Size numbers molded onto the top
  • All stoppers are 25mm long
  • Aligned with GLOBE protocols
Designed to stay pliable over long period of time, these stoppers can withstand temperatures from –25 to 70°C (–13 to 158°F). Ideal for use with nonaggressive or concentrated solutions.
 : Item contains latex, which is known to cause allergic reactions in some people. Use with caution.
Product Family Options
  • Désignation
    Taille
  • Assorted Rubber Stoppers, 1 lb.
    1 to 6
     : -
  • Assorted Rubber Stoppers, 1 lb.
    1 to 6
    Pack of 1
    Pack of 1 - $77.65
    $77.65
    Case of 5
    Case of 5 - $375.10
    $375.10
    Pack of 1
       $77.65
    Case of 5
       $375.10
     : ZM29SWMXAST1
  • Assorted Rubber Stoppers, 2 lbs.
    00 to 8
    Pack of 1
    Pack of 1 - $147.25
    $147.25
    Pack of 1
       $147.25
     : ZM29VWRASST2
  • 00 to 8
     : -
  • 1 to 8
     : -