Rubber Magdeburg Hemispheres
Catalogue # 470005-608
Fournisseur: JET RUBBER COMPANY
Spécifications
- Description:Rubber Magdeburg Hemispheres, 3.25" (8.5 cm)
- Cat. no.:470005-608
- Supplier No.:MR.GRABBER
Spécifications
À propos de cet article
Demonstrate the force of air pressure without a vacuum pump. The edges of the two 8.5 cm rubber cups are placed together and pressed to expel the air between them. Students discover the difficulty of separating the cups by pulling and tugging on the handles - providing an effective, 'hands-on' demonstration of air pressure.