Rolling Chemical Table | Avantor
Rolling Chemical Table
Labl cart, 19", 35½×36½"
Catalog # 470203-268
Supplier:   Labconco
Fournisseur #:  8020000
Rolling Chemical Table
Catalog # 470203-268
Supplier:  Labconco
Rolling Chemical Table
Rolling Chemical Table
Labl cart, 19", 35½×36½"
Price
$1,500.00
Log in pour la disponibilité
Each
Product Details & Documents
For transporting laboratory supplies and equipment, table is made of 1" square epoxy-coated steel tubing and has two shelves of high pressure melamine laminate.
  • Rails around shelves
  • 4" hard rubber casters minimize chance of accidents
Shipping weight: 44 lbs.
Spécifications
  • Désignation:
    Labl cart
  • Hauteur:
    19"
  • W×D:
    35½×36½"