ContactUsLinkComponent
Rhodospirillum rubrum Slide
Rhodospirillum rubrum Slide
Catalog # 470180-870
Supplier:  Avantor
undefined
Rhodospirillum rubrum Slide
Catalog # 470180-870
Supplier:  Avantor
Specifications
  • Description:
    Rhodospirillum rubrum
  • Cat. no.:
    470180-870
  • Stain:
    Gram (-)
  • Supplier No.:
    WARD470180-870
  • Orientation:
    Smear
Specifications
About this item
Facultative Anerobic Proteobacterium
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Spiral-shaped rods found widely distributed in nature.