Reproductive System Slide Set
Reproductive System Slide Set, 21, Animal
470183-610
: Avantor
Sections of Major Mammalian Reproductive Organs
- Identifying features clearly distinguishable
- Rigorous quality control standards
: Set includes Testis (2 slides); Epididymis (2 slides); Vas deferens; Seminal vesicle; Prostate; Sperm types (2 slides); Penis; Uterus (3 slides); Ovary (4 slides); Oviduct (3 slides); Placenta.
- Type:Animal
- Désignation:Reproductive System Slide Set
- No. of Slides:21