Relationship Rule
Relationship rule
Catalog # 470110-714
Supplier: SI Manufacturing
Supplier #: 11340X
Jump to:
Product Details & Documents
cm/mm on one edge, dm/cm on the other. Shows 1 dm equals 100 mm.
Appropriate for ages 5 and up.
Measures up to: 30 cm, Graduations: dm, cm, mm
cm/mm on one edge, dm/cm on the other. Shows 1 dm equals 100 mm.
Appropriate for ages 5 and up.
Measures up to: 30 cm, Graduations: dm, cm, mm
Specifications
- Désignation:Relationship rule