ContactUsLinkComponent
Recyclable Plastics: Identification using Relative Density and Flammability
Recyclable Plastics: Identification using Relative Density and Flammability
Catalog # 470023-904
Supplier:  Lab-Aids
undefined
Recyclable Plastics: Identification using Relative Density and Flammability
Catalog # 470023-904
Supplier:  Lab-Aids
Supplier Number:  704
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Recyclable Plastics, 16 Lab Groups, Includes Teacher's Guide and Student Copymaster
  • Cat. no.:
    470023-904
  • Supplier No.:
    704
Specifications
About this item
Identify and Construct Unknown Polymers
  • Denity, Polymers, Enviornmental Chemistry and More
Besides having students identify polymers, this lab activity can be used as an enrichment unit on density. Students use specially designed trays to identify recyclable plastics by determining their relative density. They also test the flammability of the plastics and their physical characteristics. Students attempt to identify unknown plastics using the information they have collected. Then, using molecular model components, students construct each repeat unit (the continuous molecule that makes up the larger polymer or plastic) in the polymer structure.
Ordering information: Includes Teacher Guide with SDS, Student Worksheets and Guides, Candles in holders, Containers with Lids, Copper Wires, Density Trays, Forceps, Stock Density solutions, Unknown plastic strips package, Resin package, Plastic strip package, and Molecular model component Package.