ContactUsLinkComponent
Ward's® Chemistry Reaction In The Bag Inquiry Demonstration and Lab Activity
Catalogue # 470013-586
Fournisseur:  Avantor
undefined
Ward's® Chemistry Reaction In The Bag Inquiry Demonstration and Lab Activity
Catalogue # 470013-586
Fournisseur:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Ward's® Chemistry Reaction In The Bag Inquiry Demonstration and Lab Activity
  • Cat. no.:
    470013-586
  • Supplier No.:
    WARD470013-586
  • No. of Uses:
    60
Spécifications
À propos de cet article
A Great Exploration Activity into Many Chemistry Topics
  • Students are able to design their own experiments, try them out and make observations
  • A great exploration activity into many chemistry topics
Add two dry powders and an indicator solution to the reaction bag and observe what happens. The bag cools down and the solution changes colors, then it heats up and produces a gas. Ask students what chemical reactions are occurring in this simple, yet dynamic reaction. Then, explore with them what combinations of reactants result in each of the observations. The activity can also be used to introduce writing chemical reactions and using heat as a reactant or a product. This demonstration is a natural opening to a guided inquiry activity and a great introduction to chemical reactions and chemical equations. Contains enough materials to repeat the demonstration about 20 times, or 60 smaller student-led reactions.