Ratfish
Formalin Preserved Ratfish, 8" or larger, Uninjected, Vacuum Pack of 1
Catalog # 470001-494
Supplier:   Avantor
Ratfish
Catalog # 470001-494
Supplier:  Avantor
Ratfish
Ratfish
Formalin Preserved Ratfish, 8" or larger, Uninjected, Vacuum Pack of 1
Price
$29.95
Log in for availability
Each
Product Details & Documents
Formalin preserved ratfish. Available in vacuum packs.
  • Ichthiology
  • High quality preservation
Hydrolagus colliei
Silvery and iridescent, this fish features prominent lateral line canals.
Size: 8" or more.
Specifications
  • Désignation:
    Formalin Preserved Ratfish
  • Taille:
    8" or larger
  • Injection Type:
    Uninjected
  • Packaging:
    Vacuum Pack of 1
Accessories
Product Information
  • Catalog #
    Grade
    Size
    Availability
    Price
  • WARDSafe™
    WARDSafe™
    470233-546
    Laboratory
    19 L
    Log in for availability
    Each (19l) - $345.85