Rapitest Soil Test Kit | Avantor
Rapitest Soil Test Kit
Supplier:   Luster Leaf
Rapitest Soil Test Kit
Supplier:  Luster Leaf
Certificates
Rapitest Soil Test Kit
Rapitest Soil Test Kit
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Perform qualitative soil tests without the use of chemical reagents and test tubes.
  • No hazardous compounds required for testing
  • Colorimetric determination of soil components
  • Instructions included
This kit lets students use a simple three-step process involving water, tablets and color comparators. To test soil, simply mix it with water and let it settle. Then pour the liquid off into the color comparator, add a tablet and compare the liquid with the color film chart for a test reading.
470104-846 includes 10 pH soil test kits and fully instructions.
470175-090 includes 10 tests each for pH, nitrogen, phosphorus and potash and full instructions.
Product Family Options
Informations sur le produit
  • Désignation
    No. of Tests
    Availability
    Price
  • Test kit, soil pH, Rapitest
    10
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $8.30
    $8.30
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $85.50
    $85.50
    Each
       $8.30
    Case of 12
       $85.50
    Supplier Number: 1612
  • Test kit, soil pH N, P, K, Rapitest
    40
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $35.70
    $35.70
    Log in pour la disponibilité
    Case of 6
    Case of 6 - $210.00
    $210.00
    Each
       $35.70
    Case of 6
       $210.00
    Supplier Number: 1601