Rapitest™ Moisture Meter | Avantor
Rapitest™ Moisture Meter
Rapitest™ Moisture Meter
Catalog # 470145-094
Supplier:   Luster Leaf
Fournisseur #:  1820
Rapitest™ Moisture Meter
Catalog # 470145-094
Supplier:  Luster Leaf
Rapitest™ Moisture Meter
Rapitest™ Moisture Meter
Rapitest™ Moisture Meter
Price
$73.00
Log in pour la disponibilité
Each Case of 6
Product Details & Documents
This handy meter measures soil moisture at the root level to prevent over or underwatering. No batteries are needed, so there is nothing to replace or wear out. It includes an instruction booklet, watering guide, and special needs information for 15o plants that correspond to meter readings. Range: 0 (dry) to 10 (wet). Size: 4"L x 2 1/2"W x 1 1/2"H. Probe: 4 1/4"L.
Spécifications
  • Désignation:
    Rapitest™ Moisture Meter