Raphanus, Root Tip Slide
Raphanus, Root Tip, Whole mount, Fuchsin and Fast Green
Catalog # 470181-386
Supplier: Avantor
Jump to:
Product Details & Documents
Illustrates how Plants Increase Surface Area to Absorb Nutrients
- Identifying features clearly distinguishable
- Rigorous quality control standards
Radish. Young root tip showing root hairs and cap.
Spécifications
- Désignation:Raphanus, Root Tip
- Stain:Fuchsin and Fast Green
- Orientation:Whole mount