Ward's® Live Ramshorn Snail
Ramshorn Snail, ¼ - ½"
Catalog # 470176-258
Supplier: WATERSCAPES AQUATIC NURSERY
: 874161
Jump to:
Product Details & Documents
Planorbids, mixed genera. Compare these attractive, spiral-shelled, herbivorous freshwater specimens with other snails. They will eat aquarium plants
- Herbivorous freshwater snails that will interact well with other snails.
- Will help clean your aquarium by eating debris and algae
: These products contain living or perishable materials
: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Spécifications
- Désignation:Ramshorn Snail
- Body Size:¼ - ½"