Radiation Lab Activity
Supplier:   Science First
Radiation Lab Activity
Supplier:  Science First
Certificates
Radiation Lab Activity
1 Options Available Below
Product Details & Documents
Students investigate the absorption and radiation of energy on different surfaces.
  • Activity Includes Light Source
  • Two Experiments in One Activity
  • Students Research Absorption and Radiation of Energy
They then turn on the light at 10 cm distance and observe how temperatures respond differently in the black can and silver can.
In the reverse experiment, the rates of cooling can be determined after students turn off the light.
 : Each kit includes a 16 oz. black metal and a 16 oz. shiny metal can, two insulating lids, and two other thermometers (-20° to 50°C). The single kit also includes one 120VAC clamp lamp with a reflector and 100 watt light bulb; the class kit includes four lamps.
Product Family Options
Activity
Informations sur le produit
  • Désignation
    No. of Lab Groups
    Availability
    Price
  • Radiation Lab Activity, Materials for Eight Lab Groups
    8
    Discontinued
    Supplier Number: -
  • Radiation Lab Activity, Materials for One Lab Group
    1
    Discontinued
    Supplier Number: -
  • Lab Activity, Radiation Kit, ESCP, For 15 Student Groups
    15
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 1
    Kit of 1 - $1,044.40
    $1,044.40
    Kit of 1
       $1,044.40
    Supplier Number: 16226
Lamp for Activity
Informations sur le produit
  • Catalog #
    Désignation
    Availability
    Price
  • Clamp lamp with reflector
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $45.95
    $45.95
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $45.95
    $45.95
    Each
       $45.95
    Case of 12
       $45.95
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Clamp lamp with reflector
    Clamp lamp with reflector
    470004-224
    Log in pour la disponibilité
    Each - $45.95