Boreal Science Quieting Solution | Avantor
Boreal Science Quieting Solution
Supplier:   Ward's Science
Boreal Science Quieting Solution
Supplier:  Ward's Science
Certificates
Boreal Science Quieting Solution
Boreal Science Quieting Solution
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Use this solution to slow down and relax microorganisms such as Paramecium, Euglena and Stentor. The ready-to-use dropper bottle makes it easy to store and transfer quieting liquid. Includes a dropper.
Please Note: This product is designed for educational and teaching laboratories and no certificate of analysis is available.
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Boreal Quieting Solution, 30 mL, For Protozoan Slowing
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $14.75
    $14.75
    Each
       $14.75
    Supplier Number: BB0161-30M
  • Boreal Quieting Solution, 100 mL, For Protozoan Slowing
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $19.50
    $19.50
    Each
       $19.50
    Supplier Number: BB0161-100