ContactUsLinkComponent
Ward's® Quantifying Enzyme Activity with Amylase Lab Activity
Catalogue # 470024-116
Fournisseur:  Avantor
undefined
Ward's® Quantifying Enzyme Activity with Amylase Lab Activity
Catalogue # 470024-116
Fournisseur:  Avantor
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Spécifications
  • Description:
    Quantifying Enzyme Activity with Amylase Kit
  • No. of Lab Groups:
    15
  • Cat. no.:
    470024-116
Spécifications
À propos de cet article
Unique Method Allows Easy Observation of Starch Breakdown
  • Vividly demonstrates enzyme catalysis
  • Materials for 15 lab groups
  • Time Required: ~60 minutes
In this engaging activity, students will vary the concentration of amylase, which is an enzyme that breaks down starch in order to quantify its effects. A vivid, quantifiable result is provided via the use of a starch indicator. It includes enough materials for 15 setups, a teacher’s guide, and student study guide copy-master.
Ordering information: Coupon is included for redemption of perishable materials. Redeem by mail, phone, fax, or email.