Quality Precleaned Glass Microscope Slides
Quality precleaned glass slides, 0.9 - 1.0 mm, 75×25 mm (3×1")
Catalog # 470145-790
Supplier: C & A Scientific
Fournisseur #: 9101-E
Jump to:
Product Details & Documents
Dust-free and ready to use.
- Quality slide making supplies
- All the work is done for you
Distortion-free glass is cut, ground, and polished to precision standards. Slides are chemically washed, rinsed, and dried.
Spécifications
- Désignation:Quality precleaned glass slides
- L×l:75×25 mm (3×1")
- Épaisseur:0.9 - 1.0 mm