Ward's® Pyroxene Thin Section Set
Pyroxene Set, 11
Catalog # 470024-860
Supplier: Avantor
Jump to:
Product Details & Documents
Great value for the microscopic study of Pyroxene minerals
Specially Selected by Wards® staff Geologists
- 11 Thin Sections
- Includes Key for Easy Identification
Specially selected thin sections of important rock forming minerals. Set may contain more than one example of the same mineral to depict variations in occurrence and distinct mineral associations.
Slides (11): acmite augite (2) diopside, enstatite, fassaite, hypersthene, hedenbergite, johannsenite, pigeonite, spodumene.
Ordering information: Thin sections subject to changed based on availability.
Spécifications
- Désignation:Pyroxene Set
- No. of Slides:11