Pyroglas™ Fire Blanket with Wall Case
72" L x 60" W
Catalog # 470122-240
Supplier: Cotton Goods
Fournisseur #: FR2517
Jump to:
Product Details & Documents
For smothering fires.
- Clean, soft surface
- Insulates, repels water, and is impervious to most acids, alkalis, and solvents
Blanket is stored in a metal, wall-mounted fire red container which opens easily at the bottom for quick removal. Container dimensions: 18"H×7"Ø
Spécifications
- Dimensions:72" L x 60" W