PYREX® Volumetric Pipettes, Color-Coded, Class A | Avantor
PYREX® Volumetric Pipettes, Color-Coded, Class A
   Options durables disponibles
Supplier:   Corning
PYREX® Volumetric Pipettes, Color-Coded, Class A
Supplier:  Corning
   Options durables disponibles
Certificates
PYREX® Volumetric Pipettes, Color-Coded, Class A
PYREX® Volumetric Pipettes, Color-Coded, Class A
4 Options Available Below
Product Details & Documents
These pipettes are made of high-grade borosilicate glass.
  • Reusable, rugged construction
  • Color-coded
  • Colored enamel graduations and markings
Ruggedly constructed and properly proportioned, pipettes have accuracy tolerances to meet or exceed the strict requirements for Class A Volumetric Ware as indicated by ASTM E969.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 470211-598
  • 470211-606
  • 470211-600
  • 470211-604
  • 470211-608
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Capacité
    Availability
    Price
  • Pyrex® transfer pipettes (volumetric pipettes), cap., each
    1 ml
    Discontinued
    Supplier Number: 7100-1
    Spécifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    470211-598
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
      Low Manufacturing Impact
  • Pyrex® transfer pipettes (volumetric pipettes), cap., each
    5 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $28.00
    $28.00
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $336.00
    $336.00
    Each
       $28.00
    Case of 12
       $336.00
    Supplier Number: 7100-5
    Spécifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    470211-606
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
      Low Manufacturing Impact
  • Pyrex® transfer pipettes (volumetric pipettes), cap., each
    10 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $30.60
    $30.60
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $367.20
    $367.20
    Each
       $30.60
    Case of 12
       $367.20
    Supplier Number: 7100-10
    Spécifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    470211-600
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
      Low Manufacturing Impact
  • Pyrex® transfer pipettes (volumetric pipettes), cap., each
    25 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $47.80
    $47.80
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $573.60
    $573.60
    Each
       $47.80
    Case of 12
       $573.60
    Supplier Number: 7100-25
    Spécifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    470211-604
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
      Low Manufacturing Impact
  • Pyrex® transfer pipettes (volumetric pipettes), cap., each
    50 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $88.60
    $88.60
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $1,063.20
    $1,063.20
    Each
       $88.60
    Case of 12
       $1,063.20
    Supplier Number: 7100-50
    Spécifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    470211-608
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
      Low Manufacturing Impact