Order Entry
Canada
ContactUsLinkComponent
PYREX® Rimless Culture Tubes
  470211-678
 :  Corning
undefined
PYREX® Rimless Culture Tubes
  470211-678
 :  Corning
 :  9820-6

 

  • Description:
    Pyrex® Rimless Culture Tubes, OD x L, mm: 6 x 50, Cap., mL: 0.5
  • Cat. no.:
    470211-678
  • Supplier No.:
    9820-6

 

 

Made of PYREX® borosilicate glass, rimless.

  • Very good chemical resistance
  • Very good temperature resistance
  • Ends are fire-polished

Culture tubes have walls and bottom of a uniform thickness. All tubes feature a white marking spot, with the exception of 6 and 22 mm tubes.

Tube 5 ml can be used for Kahn tests. Tube 9 ml can be used for Wasserman tests.