PYREX® Long Stem Funnels, Corning
PYREX® Long Stem Funnels, For Paper Dia 11 cm, 65 mm
Supplier: Corning
Supplier #: 6140-65
Jump to:
Product Details & Documents
Corning No. 6140 Strong beaded edge. 60° angle. Stems of heavy wall tubing approximately 150 mm (6") long. Package of 12.
Specifications
- Désignation:PYREX® Long Stem Funnels, For Paper Dia 11 cm
- Top I.D.:65 mm