PYREX® Liebig Condenser
Pyrex® Liebig Condenser, Jacket: 41 mm x 200 mm, Overall Length: 345 mm
Supplier: Corning
Supplier #: 2340-200
Jump to:
Product Details & Documents
A fine borosilicate condenser with the inner tube sealed in.
The large-capacity water jacket ensures efficient cooling. The adapter end will accept a No. 3 rubber stopper.
Specifications
- Désignation:Pyrex® Liebig Condenser, Jacket: 41 mm x 200 mm, Overall Length: 345 mm