Pyrex® Single-Scale Precision Cylinders
Supplier: Corning
À propos de cet article
Borosilicate glass.
- Calibrated to contain (TC In ) at 20 °C (68 °F)
- Hexagonal base with graduations in white enamel
- 10 ml cylinder has a funnel top
Use the 100 ml cylinder for ASTM E133, E1272, D86, D216, and D447.
Delivery information: Supplied with bumper guards for cylinders of capacity 25 to 500 ml.
6 Options Available Below
Product Details & Documents
Borosilicate glass.
- Calibrated to contain (TC In ) at 20 °C (68 °F)
- Hexagonal base with graduations in white enamel
- 10 ml cylinder has a funnel top
Use the 100 ml cylinder for ASTM E133, E1272, D86, D216, and D447.
Delivery information: Supplied with bumper guards for cylinders of capacity 25 to 500 ml.
Specifications for Product Family
Spécifications
- Catalog No:
- 470202-082
- 470202-088
- 470202-092
- 470202-084
- 470211-424
- 470211-426
- Form:
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
Spécifications
Product Family Options
Informations sur le produit
- CapacitéIntervalle de graduationAvailabilityPrice
- 10 ml0.1 mlEach$23.70Case of 24$569.20Supplier Number: 3022-10Spécifications:Product available on GSA Advantage®
More
Specifications
Cat. No.470202-082FormTallEnvironmentally PreferableEnvironmental certificationProduct available on GSA Advantage® - 25 ml0.5 mlEach$52.40Case of 18$943.20Supplier Number: 3022-25Spécifications:
More
Specifications
Cat. No.470202-088FormTallEnvironmentally PreferableEnvironmental certification - 50 ml1 mlEach$54.20Case of 18$975.60Supplier Number: 3022-50Spécifications:Product available on GSA Advantage®
Low Manufacturing Impact More
Specifications
Cat. No.470202-092FormTallEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - Monterrey - 100 ml1 mlEach$64.60Case of 12$775.20Supplier Number: 3022-100Spécifications:Product available on GSA Advantage®
Low Manufacturing Impact More
Specifications
Cat. No.470202-084FormTallEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - Monterrey - 250 ml2 mlEach$59.90Case of 12$718.85Supplier Number: 3022-250Spécifications:
Low Manufacturing Impact More
Specifications
Cat. No.470211-424FormTallEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - MonterreyLow Manufacturing Impact - 500 ml5 mlEach$125.00Case of 8$1,000.00Supplier Number: 3022-500Spécifications:
More
Specifications
Cat. No.470211-426FormTallEnvironmentally PreferableEnvironmental certification