PYREX® Ignition Tubes
PYREX® Ignition Tubes
Catalog # 470211-684
Supplier:   Corning
Supplier #:  9860-16
PYREX® Ignition Tubes
Catalog # 470211-684
Supplier:  Corning
PYREX® Ignition Tubes
PYREX® Ignition Tubes
Price
$400.00
Log in for availability
Pack of 72 Case of 288
Product Details & Documents
These PYREX® rimless ignition tubes are made from No. 7740 glass.
  • Very good chemical resistance
  • Good temperature resistance
These ignition tubes are satisfactory for most applications encountered in school work where temperatures above 600 °C are not required. These rimless tubes perform well when continually used at temperatures up to 500 °C, or for short periods to 600 °C.