ContactUsLinkComponent
PYREX® Heavy-Duty-Grade Beakers
PYREX® Heavy-Duty-Grade Beakers
Catalog #
Supplier:  Corning
Supplier #:
undefined
PYREX® Heavy-Duty-Grade Beakers
Catalog #
Supplier:  Corning
Supplier Number:  1003-150
Supplier #:
Select an Option
Select an Option
Select an Option
Select an Option
Pyrex® Heavy Duty Beakers
About this item
Made of PYREX® borosilicate glass.
  • Very good chemical resistance
  • Good temperature resistance
  • With extra large marking spot
Heavy-duty beakers specifically designed to offer the best mechanical strength under harsh conditions, such as mechanised washing operations. Sizes 250 ml and larger have a double graduated metric scale to indicate approximate content.
Certifications: Meet ASTM E960 specifications.
Frequently Bought Together