PYREX® Filtering Flasks | Avantor
PYREX® Filtering Flasks
   Options durables disponibles
Supplier:   Corning
PYREX® Filtering Flasks
Supplier:  Corning
   Options durables disponibles
Certificates
PYREX® Filtering Flasks
3 Options Available Below
Product Details & Documents
PYREX® borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Flasks have a beaded top that provides an excellent fit for rubber stoppers. Graduated to show approximate volume.
The heavy-duty rim flange is ideal for supporting Buchner funnels. It includes a side arm for attaching rubber tubing. Extra-large marking spot for easy identifications.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 470211-504
  • 470211-506
  • 470211-502
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
Spécifications
Product Family Options
Informations sur le produit
  • Capacité
    Availability
    Price
  • 250 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 6
    Pack of 6 - $271.00
    $271.00
    Log in pour la disponibilité
    Case of 18
    Case of 18 - $813.00
    $813.00
    Pack of 6
       $271.00
    Case of 18
       $813.00
    Supplier Number: 5340-250
    Spécifications:
    More
    Specifications
    Cat. No.
    470211-504
    Environmentally Preferable
    Environmental certification
  • 500 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 6
    Pack of 6 - $379.00
    $379.00
    Log in pour la disponibilité
    Case of 18
    Case of 18 - $1,137.00
    $1,137.00
    Pack of 6
       $379.00
    Case of 18
       $1,137.00
    Supplier Number: 5340-500
    Spécifications:
    More
    Specifications
    Cat. No.
    470211-506
    Environmentally Preferable
    Environmental certification
  • 1000 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $118.00
    $118.00
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $1,416.00
    $1,416.00
    Each
       $118.00
    Case of 12
       $1,416.00
    Supplier Number: 5340-1L
    Spécifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    470211-502
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
      Low Manufacturing Impact