Pyrex® Erlenmeyer Flasks
 :   Corning
Pyrex® Erlenmeyer Flasks
 :  Corning
Pyrex® Erlenmeyer Flasks
Pyrex® Erlenmeyer Flasks
PYREX® borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Heavy duty rim flasks have approximate graduations in durable white enamel. Extra large marking space. Heavy, durable top tooling.
Product Family Options
  • Capacité
    Intervalle de graduation
    Plage de graduation
    Stopper No.
  • 25 ml
    5 ml
    10 - 25 ml
    0
    Pack of 12
    Pack of 12 - $150.00
    $150.00
    Case of 48
    Case of 48 - $600.00
    $600.00
    Pack of 12
       $150.00
    Case of 48
       $600.00
     : 4980-25
  • 50 ml
    10 ml
    20 - 50 ml
    1
    Pack of 12
    Pack of 12 - $146.00
    $146.00
    Case of 48
    Case of 48 - $584.00
    $584.00
    Pack of 12
       $146.00
    Case of 48
       $584.00
     : 4980-50
  • 125 ml
    25 ml
    50 - 125 ml
    5
    Pack of 12
    Pack of 12 - $138.00
    $138.00
    Case of 48
    Case of 48 - $552.00
    $552.00
    Pack of 12
       $138.00
    Case of 48
       $552.00
     : 4980-125
  • 250 ml
    25 ml
    50 - 200 ml
    6
    Pack of 12
    Pack of 12 - $132.00
    $132.00
    Case of 48
    Case of 48 - $528.00
    $528.00
    Pack of 12
       $132.00
    Case of 48
       $528.00
     : 4980-250
  • 500 ml
    50 ml
    100 - 500 ml
    7
    Pack of 6
    Pack of 6 - $82.40
    $82.40
    Case of 36
    Case of 36 - $494.40
    $494.40
    Pack of 6
       $82.40
    Case of 36
       $494.40
     : 4980-500
  • 1000 ml
    50 ml
    250 - 1000 ml
    9
    Each
    Each - $22.30
    $22.30
    Case of 24
    Case of 24 - $535.20
    $535.20
    Each
       $22.30
    Case of 24
       $535.20
     : 4980-1L
  • 2000 ml
    200 ml
    600 - 1800 ml
    10
    Each
    Each - $52.60
    $52.60
    Case of 8
    Case of 8 - $420.80
    $420.80
    Each
       $52.60
    Case of 8
       $420.80
     : 4980-2L