PYREX® Erlenmeyer Flask with Screw Cap
PYREX® Erlenmeyer Flask with Screw Cap
Supplier:   Corning
PYREX® Erlenmeyer Flask with Screw Cap
Supplier:  Corning
Certificates
PYREX® Erlenmeyer Flask with Screw Cap
PYREX® Erlenmeyer Flask with Screw Cap
2 Options Available Below
Product Details & Documents
Made of PYREX® borosilicate glass, with phenolic screw caps.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Graduated to show approximate capacity
  • With marking spot
Erlenmeyer flasks with phenolic screw caps are ideal for use as a shaker flask or for mixing, media prep and storage needs. Impact resistant caps are equipped with cemented-in, inert white rubber liners which are resistant to steam sterilization. Their uniform wall thickness provides the proper balance between mechanical strength and thermal shock resistance.
Product Family Options
Informations sur le produit
  • Capacité
    Availability
    Price
  • 500 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $53.40
    $53.40
    Log in pour la disponibilité
    Pack of 6
    Pack of 6 - $320.40
    $320.40
    Log in pour la disponibilité
    Case of 24
    Case of 24 - $1,281.60
    $1,281.60
    Each
       $53.40
    Pack of 6
       $320.40
    Case of 24
       $1,281.60
    Supplier Number: 4985-500
  • 1000 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $72.10
    $72.10
    Log in pour la disponibilité
    Pack of 6
    Pack of 6 - $432.60
    $432.60
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $865.20
    $865.20
    Each
       $72.10
    Pack of 6
       $432.60
    Case of 12
       $865.20
    Supplier Number: 4985-1L