PYREX® Burets
PYREX® Burets
   Options durables disponibles
Supplier:   Corning
PYREX® Burets
Supplier:  Corning
   Options durables disponibles
Certificates
PYREX® Burets
3 Options Available Below
Product Details & Documents
Educational grade Pyrex burets.
  • Heavy duty
  • Transparent
Supplied with self-lubricating, leakproof PMP stopcock. This buret is unaffected by most dilute aqueous solutions, except ammonia and HF. It is not recommended for use with ethanol or organic solvents. The clear white markings make dark liquids easier to measure the the Teflon stopcock is less likely to freeze up.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 470211-386
  • 470211-390
  • 470146-252
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Capacité
    Intervalle de graduation
    Availability
    Price
  • Geissler Buret
    50 ml
    0.1 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $209.00
    $209.00
    Log in pour la disponibilité
    Case of 6
    Case of 6 - $1,254.00
    $1,254.00
    Each
       $209.00
    Case of 6
       $1,254.00
    Supplier Number: 2116-50
    Spécifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    470211-386
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
      Low Manufacturing Impact
  • Geissler Buret Locking Stopcock
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $163.00
    $163.00
    Each
       $163.00
    Supplier Number: 2116-LSO
    Spécifications:
    More
    Specifications
    Cat. No.
    470211-390
    Environmentally Preferable
    Environmental certification
  • Geissler buret replacement tip, pack of 12, six grinded tips and six clear (ungrinded) tipss
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 12
    Pack of 12 - $197.00
    $197.00
    Pack of 12
       $197.00
    Supplier Number: 2116-GTO2
    Spécifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    470146-252
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
      Low Manufacturing Impact