Fundamentals of Physics Pulley System
Catalogue # 470014-338
Fournisseur: Wards
Spécifications
- Description:FUNdamentals of Physics Pulley System
- Cat. no.:470014-338
- Supplier No.:RPL08
Spécifications
À propos de cet article
Discover the Mechanical Advantage of Different Pulley Combinations
- Multiple pulleys in one comprehensive device, this system allows students to experiment with many variations of string length, force, and more
- Requires the Fun stand, sold separately
Demonstrate that the length of string pulled increases with the number of pulleys used. Also calculate the work conserved between input and output work when the amount of force put into the system decreases with the more pulleys students add.