Pulley Demonstration Set | Avantor
Pulley Demonstration Set
Pulley Demo, Student, Set
Catalog # 470016-048
Supplier:   Wards
Fournisseur #:  PH0305
Pulley Demonstration Set
Catalog # 470016-048
Supplier:  Wards
Pulley Demonstration Set
Pulley Demonstration Set
Pulley Demo, Student, Set
Price
$560.30
Log in pour la disponibilité
Each
Product Details & Documents
A simpler version of the Advanced Pulley Demonstration Set, this model includes eight single pulleys and two double sheave pulleys in addition to EISCO ball bearing frictionless pulley.
  • Economical
  • Ball-Bearing, Frictionless Pulley
  • Wooden Base
  • Suitable for Introductory Pulley Demonstrations
A wooden base supports one vertical rod with attachments to a horizontal rod at top.
This set has everything needed to conduct a series of introductory pulley demonstrations.
Ordering information: This kit includes hooked masses, collars with hooks, clamps, and cord.
Spécifications
  • Désignation:
    Pulley Demo, Student, Set