Pulley Cords | Avantor
Pulley Cords
Supplier:   Arcor Electronics
Pulley Cords
Supplier:  Arcor Electronics
Certificates
Pulley Cords
5 Options Available Below
Product Details & Documents
Ideal for small pulley systems, cord is available in a variety of colors, strengths, and lengths.
  • Ideal for pulley systems, hanging masses, and wave demonstrations
  • Nylon and Dacron are extremely durable
  • Ends may be heat fused to prevent fraying
  • Premium Dacron cords resists stretching and fraying
Every physics classroom needs good-quality string - whether it's for an Atwood's machine, pulling friction blocks, or hanging pendulums. Teachers need string that is easy to knot and won't unravel after it is cut. These products have been selected for durability and strength, and come on a convenient spool for easy dispensing and storage.
Product Family Options
Informations sur le produit
  • Désignation
    Color
    Longueur
    Availability
    Price
  • Braided Nylon pulley cord, tensile strength 15 lbs. test
    Black
    45 m (50 yds.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $35.80
    $35.80
    Each
       $35.80
    Supplier Number: 6CB-15
  • Braided Dacron pulley cord, tensile strength 30 lbs. test
    White
    45 m (50 yds.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $19.50
    $19.50
    Each
       $19.50
    Supplier Number: 50CD-130
  • Premium Dacron pulley cord, tensile strength 30 lbs. test
    White/green
    45 m (50 yds.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $21.05
    $21.05
    Each
       $21.05
    Supplier Number: 30LB50YARD
  • Heavy duty Dacron pulley cord, tensile strength 135 lbs. test
    Orange
    9 m (10 yds.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $14.90
    $14.90
    Each
       $14.90
    Supplier Number: 10PB-135
  • Braided Nylon pulley cord, tensile strength 200 lbs. test
    White
    9 m (10 yds.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $31.00
    $31.00
    Each
       $31.00
    Supplier Number: SDX16165
  • Pulley cord
    Discontinued
    Supplier Number: -
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Support base steel for braided nylon pulley cord, tensile strength 15 lbs. test, height: 23 mm, base dimension: 5×7", thread size: ³/₈-16
    Support base steel for braided nylon pulley cord, tensile strength 15 lbs. test, height: 23 mm, base dimension: 5×7", thread size: ³/₈-16
    470123-472
    Log in pour la disponibilité
    Each - $36.65