Pulley Bench Clamp Fitting
Pulley Bench Clamp Fitting
470120-950
: Wards
: PH0296 A
A Steady Hold
- Designed for clamping to the edge of a bench, table, and more, this pulley overhangs the edge in a vertical plane.
The device consists of a 50mm diameter pulley carried on a two part aluminum frame. Maximum clamp opening is 25mm. Overall frame length is 110mm.
- Désignation:Pulley Bench Clamp Fitting