PTC Taste Test Paper
470123-120
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Description:PTC Taste Test Paper
- Cat. no.:470123-120
- Supplier No.:165-12V-100
Genetic Test Paper
- Pocket-Sized, Lightweight Vial
- 100 test strips per vial
Teach students about dominant and recessive genes with PTC Taste Test Paper. The ability to taste bitterness of the test paper is usually considered as a dominant genetic trait, whereas the inability to taste the paper is considered a recessive trait. Seven out of ten people are able to taste PTC.