Psychrometer Kit Set/15°C | Avantor
Psychrometer Kit Set/15°C
Psychrometer Kit (°C), Set of 15
Catalog # 470023-946
Supplier:   Science First
Fournisseur #:  16221
Psychrometer Kit Set/15°C
Catalog # 470023-946
Supplier:  Science First
Psychrometer Kit Set/15°C
Psychrometer Kit Set/15°C
Psychrometer Kit (°C), Set of 15
Liquidation
$121.05
Vente
Valable jusqu'au 31/12/2026
Log in pour la disponibilité
Kit of 15
Product Details & Documents
Measure humidity and temperature with this psychrometer kit.
  • Includes thermometer and hygrometer
    Ideal for environmental and weather studies
    –40 to +50 °C
    6½×¾" size
Explore Weather Science with the Psychrometer Kit Perfect for hands-on learning, this kit enables students to measure both temperature and humidity—key elements in understanding weather and atmospheric conditions. Every kit features two vinyl-backed alcohol thermometers mounted on durable plastic handles, along with a wick, rubber band, and a vapor pressure/humidity chart for accurate readings.
Delivery information: Kit includes materials to make 15 the psychrometers: 2 thermometers, a handle, wet bulb wick, and psychrometric chart.
Spécifications
  • Désignation:
    Psychrometer Kit (°C), Set of 15