Protein Folding Study Activity Kit
Protein Folding Study Activity Kit
470149-920
: BE-411
Demonstrate Principles of Protein Denaturation and Refolding
Protein unfolding and folding, a reversible process, is analyzed using a light emitting protein specifically prepared for this study and a protein electrophoresis technique. The kit allows students to see when a light emitting protein is denatured (unfolded) as it loses its property to emit light. Materials supplied for six groups of up to five students each.
- Désignation:Protein Folding Study Activity Kit