Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Description:Properties of Acids & Bases Experiment Lab Activity
- Cat. no.:470007-024
- Includes:Teacher ‘s Guide and Student Copymasters
- Supplier No.:82
- No. of Uses:24
This Applied Science Concepts Kit provides a completely safe and convenient method for students to test and observe the characteristic properties of acids and bases.
- Seven Activities Included
- Students Form a Salt with Neutralization Experiment
- No Need for Glassware
- Semi-Micro Environment in Any Classroom
- Safe and Convenient Experiments
Activities incorporate the LAB-AIDS® Chemplates®. This unique lab tool eliminates the need for glassware and provides a semi-micro environment in any classroom.
Students perform seven activities such as reacting metal and carbonates with acids, producing, collecting, and testing carbon dioxide and hydrogen in safe quantities, observing the effects of acid and bases on indicator dyes, measuring the concentration of acids by titration of an “unknown,” and performing a neutralization experiment.