Project STAR Spectrometer | Avantor
Project STAR Spectrometer
Project STAR Spectrometer
Catalog # 470003-848
Supplier:   Science First
Fournisseur #:  614-0210
Project STAR Spectrometer
Catalog # 470003-848
Supplier:  Science First
Project STAR Spectrometer
Project STAR Spectrometer
Project STAR Spectrometer
Liquidation
$50.75
Vente
Valable jusqu'au 31/12/2026
Log in pour la disponibilité
Each
Product Details & Documents
View Spectra with an Accuracy of ±1 Nanometer
Examine the spectra of the Sun and stars, and those produced by minerals in flame tests. It features a high-quality grating and stepped slit for a clear, precise, spectrum. The sturdy plastic construction is labeled with both electron volts and nanometers, as well as a chart of common spectral lines. An instruction manual is included. Size: 12"L x 8 1/2"W.
Spécifications
  • Désignation:
    Project STAR Spectrometer