Precision Test Strips
Precision Test Strips
Precision Test Strips, Test: Nitrate, Range, ppm: 0 - 250, Color Chart Concentrations: 0, 10, 25, 50, 100, 250, 500, 0 - 250 ppm
Catalog # 470123-128
Supplier #:  NAT-1V-50
Precision Test Strips
Catalog # 470123-128
CAS Number:  
Precision Test Strips
Precision Test Strips
Precision Test Strips, Test: Nitrate, Range, ppm: 0 - 250, Color Chart Concentrations: 0, 10, 25, 50, 100, 250, 500, 0 - 250 ppm
CAS Number:  
Price
$28.20
Log in for availability
Pack of 50
Product Details & Documents
Precision Test Strips quickly test for low levels of nitrite and chlorine.
  • Fast Tests
  • Pool Test Strip Measures pH, free, and Total Chlorine
  • Aquarium Test Strip Measures pH and Chlorine Levels
  • Fifty Test Strips in One Package
  • Test Strips for Nitrite and Chlorine
After immersing the strip in the solution to be measured, the concentration is measured by comparing the strip’s color to a color chart on the package.
The Pool Test Strip is three tests in one—pH, free, and total chlorine. The aquarium test strip simultaneously measures pH and chlorine levels.
Packaging: Each package includes 50 test strips in a resealable package.
Specifications
Specifications
  • Désignation:
    Precision Test Strips, Test: Nitrate, Range, ppm: 0 - 250, Color Chart Concentrations: 0, 10, 25, 50, 100, 250, 500
  • Range:
    0 - 250 ppm