Precision Gyroscope
Precision gyroscope, 7 cm (2.75"), 68 g
Catalog # 470004-966
Supplier: Tedco
Supplier #: 30000
Jump to:
Product Details & Documents
This gyroscope will spin for several minutes.
- Ready to use right out of the box
- Unparalleled ease-of-use and performance
- Use string or T-handle zip starter to initiate spinning
- Simple and effective tool to teach kids about physics
This is the most precision designed toy gyroscope on the market. Tiny ball bearings and near perfect balance keep the gyroscope spinning for several minutes. The included T-handle zip starter strap drastically improves ease-of-use. Durable enough for classroom demonstration.
Delivery information: Includes precision gyroscope, pedestal, string and T-handle zip starter.
Specifications
- Désignation:Precision gyroscope
- Weight:68 g
- Diameter:7 cm (2.75")