Powder-Free Disposable Nitrile Gloves
Supplier: Dival Safety & Supplies
À propos de cet article
These powder-free nitrile gloves are perfect for use in the laboratory or classroom.
- Latex-allergy safe
- Perfect for student use
- Cuff for added protection
- Blue
- Powder-free
They are an economical, latex-free choice for student safety and feature a cuff for extra protection.
The blue color is easy to identify in the laboratory.
3 Options Available Below
Product Details & Documents
These powder-free nitrile gloves are perfect for use in the laboratory or classroom.
- Latex-allergy safe
- Perfect for student use
- Cuff for added protection
- Blue
- Powder-free
They are an economical, latex-free choice for student safety and feature a cuff for extra protection.
The blue color is easy to identify in the laboratory.
Specifications for Product Family
Spécifications
- Catalog No:
- 470222-546
- 470356-766
- 470222-548
- 470356-752
- 470018-302
- 470356-760
- 470222-544
- 470356-754
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
Spécifications
Product Family Options
Informations sur le produit
- Taille du gantGlove ColorLongueurPackagingAvailabilityPrice
- SBlue24.1 cm (9.5")Box/100Pack of 100$10.45Case of 1000$104.70Supplier Number: GVNP-SM-1Spécifications:
More
Specifications
Cat. No.470222-546Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-free - SBlue24.1 cm (9.5")Box/100Discontinued-Supplier Number: DVPF4BLSMSpécifications:
More
Specifications
Cat. No.470356-766Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-free - MBlue24.1 cm (9.5")Box/100Pack of 100$37.65Case of 1000$340.30Supplier Number: GNPRMDSpécifications:
More
Specifications
Cat. No.470222-548Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-free - MBlue24.1 cm (9.5")Box/100Discontinued-Supplier Number: DVPF4BLMDSpécifications:
More
Specifications
Cat. No.470356-752Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-free - LBlue24.1 cm (9.5")Box/100Box of 100$50.30Case of 1000$446.30Supplier Number: GNPRLGSpécifications:
More
Specifications
Cat. No.470018-302Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-free - LBlue24.1 cm (9.5")Box/100Discontinued-Supplier Number: DVPF4BLLGSpécifications:
More
Specifications
Cat. No.470356-760Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-free - XLBlue24.1 cm (9.5")Box/100Discontinued-Supplier Number: GVNP-XL-1Spécifications:
More
Specifications
Cat. No.470222-544Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-free - XLBlue24.1 cm (9.5")Box/100Discontinued-Supplier Number: DVPF4BLXLSpécifications:
More
Specifications
Cat. No.470356-754Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-free